Product Details

SNP ID
rs77291096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:92704631 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGACCTTTCAAGCGTCACCGCTGC[A/G]TTAGTTACCTGTGAAGCGGATACGG
Phenotype
MIM: 602620
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LGMN PubMed Links

Gene Details

Gene
LGMN
Gene Name
legumain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008530.2 Intron NP_001008530.1
NM_005606.6 Intron NP_005597.3
XM_005267862.3 Intron XP_005267919.1
XM_005267863.3 Intron XP_005267920.1
XM_011536970.1 Intron XP_011535272.1
XM_017021463.1 Intron XP_016876952.1
XM_017021464.1 Intron XP_016876953.1
XM_017021465.1 Intron XP_016876954.1
XM_017021466.1 Intron XP_016876955.1
XM_017021467.1 Intron XP_016876956.1

View Full Product Details