Product Details

SNP ID
rs139957631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:107222958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTCTTTAAAAATCGTGGTTGCT[A/G]TGTTTCACGTACTAGGGTAACAATG
Phenotype
MIM: 300933
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIH1D3 PubMed Links

Gene Details

Gene
PIH1D3
Gene Name
PIH1 domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001169154.1 Intron NP_001162625.1
NM_173494.1 Intron NP_775765.1
XM_011530855.2 Intron XP_011529157.1

View Full Product Details