Product Details
- SNP ID
-
rs74962444
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:94128490 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATTTCACTGTCCTAGTGAATAAG[A/G]GACTCTAGTCGTTGGCCTTCTTGGA
- Phenotype
-
MIM: 603017
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDH17
PubMed Links
Gene Details
- Gene
- CDH17
- Gene Name
- cadherin 17
View Full Product Details