Product Details

SNP ID
hCV25600515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64767188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACCCACAGCCAGCAGACAGCCA[C/T]TACCTTTTCCTTGATGCAGGCGATA
Phenotype
MIM: 608455 MIM: 601516
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PYGM PubMed Links

Gene Details

Gene
PYGM
Gene Name
phosphorylase, glycogen, muscle
There are no transcripts associated with this gene.

Gene
SF1
Gene Name
splicing factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178030.1 Intron NP_001171501.1
NM_004630.3 Intron NP_004621.2
NM_201995.2 Intron NP_973724.1
NM_201997.2 Intron NP_973726.2
NM_201998.2 Intron NP_973727.1

View Full Product Details