Product Details

SNP ID
rs76826183
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233477719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCTTCCCCTGCTTTGCACTGGC[A/G]GCACGGCATGGTTCAGGCTCTGGGG
Phenotype
MIM: 601826 MIM: 610570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGKD PubMed Links

Gene Details

Gene
DGKD
Gene Name
diacylglycerol kinase delta
There are no transcripts associated with this gene.

Gene
USP40
Gene Name
ubiquitin specific peptidase 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018218.2 Intron NP_060688.1
XM_006712612.3 Intron XP_006712675.1
XM_011511396.2 Intron XP_011509698.1
XM_011511397.2 Intron XP_011509699.1
XM_011511398.2 Intron XP_011509700.1
XM_011511399.2 Intron XP_011509701.1
XM_011511400.2 Intron XP_011509702.1
XM_011511401.2 Intron XP_011509703.1
XM_017004426.1 Intron XP_016859915.1
XM_017004427.1 Intron XP_016859916.1
XM_017004428.1 Intron XP_016859917.1

View Full Product Details