Product Details

SNP ID
rs112256607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4247265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGAGATCTCTTCTTTGGAACC[C/T]TTCTTTCCCAGCGGCCTTCCGCGAG
Phenotype
MIM: 605816
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC94 PubMed Links

Gene Details

Gene
CCDC94
Gene Name
coiled-coil domain containing 94
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018074.4 Intron NP_060544.2
Gene
EBI3
Gene Name
Epstein-Barr virus induced 3
There are no transcripts associated with this gene.

View Full Product Details