Product Details

SNP ID
rs56250405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100526894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGAAAGCTTAGTGTTACTTGG[C/T]CAAAGAAGTGCCATCTGAGCTAAAG
Phenotype
MIM: 606615 MIM: 602140 MIM: 610258
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIF1AN PubMed Links

Gene Details

Gene
HIF1AN
Gene Name
hypoxia inducible factor 1 alpha subunit inhibitor
There are no transcripts associated with this gene.

Gene
NDUFB8
Gene Name
NADH:ubiquinone oxidoreductase subunit B8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284367.1 Intron NP_001271296.1
NM_001284368.1 Intron NP_001271297.1
NM_005004.3 Intron NP_004995.1
Gene
SEC31B
Gene Name
SEC31 homolog B, COPII coat complex component
There are no transcripts associated with this gene.

View Full Product Details