Product Details
- SNP ID
-
rs147747540
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:11920278 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGGACTTTCAGTGCTGGGTGTCTT[C/G]TGCAAATGGTGGCTGAAAGCAGGAC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA2013
PubMed Links
Gene Details
- Gene
- KIAA2013
- Gene Name
- KIAA2013
View Full Product Details