Product Details

SNP ID
rs115375801
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101588419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGACGGTAACTCGAGGGTCCCCA[C/G]GGGCTCCTTCGTTTTTTTCCCTCAG
Phenotype
MIM: 616467 MIM: 606343
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DPCD PubMed Links

Gene Details

Gene
DPCD
Gene Name
deleted in primary ciliary dyskinesia homolog (mouse)
There are no transcripts associated with this gene.

Gene
POLL
Gene Name
polymerase (DNA) lambda
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174084.1 Intron NP_001167555.1
NM_001174085.1 Intron NP_001167556.1
NM_001308382.1 Intron NP_001295311.1
NM_013274.3 Intron NP_037406.1
XM_006717775.1 Intron XP_006717838.1
XM_006717776.1 Intron XP_006717839.1
XM_006717777.1 Intron XP_006717840.1
XM_011539650.1 Intron XP_011537952.1
XM_011539651.1 Intron XP_011537953.1
XM_011539652.1 Intron XP_011537954.1
XM_011539653.1 Intron XP_011537955.1
XM_011539654.1 Intron XP_011537956.1
XM_011539655.1 Intron XP_011537957.1
XM_011539656.1 Intron XP_011537958.1
XM_011539657.1 Intron XP_011537959.1
XM_011539659.1 Intron XP_011537961.1
XM_011539660.1 Intron XP_011537962.1
XM_011539662.1 Intron XP_011537964.1
XM_011539663.1 Intron XP_011537965.1
XM_011539664.1 Intron XP_011537966.1
XM_011539665.2 Intron XP_011537967.1
XM_011539666.1 Intron XP_011537968.1
XM_011539667.1 Intron XP_011537969.1
XM_017016084.1 Intron XP_016871573.1
XM_017016085.1 Intron XP_016871574.1
XM_017016086.1 Intron XP_016871575.1
XM_017016087.1 Intron XP_016871576.1
XM_017016088.1 Intron XP_016871577.1
XM_017016089.1 Intron XP_016871578.1
XM_017016090.1 Intron XP_016871579.1
XM_017016091.1 Intron XP_016871580.1
XM_017016092.1 Intron XP_016871581.1
XM_017016093.1 Intron XP_016871582.1

View Full Product Details