Product Details

SNP ID
rs541352457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101597046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCCACCCCCCACAATGATTGCT[G/T]CATGTATTCATAGAGGAAAAAACTG
Phenotype
MIM: 616467 MIM: 606343
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DPCD PubMed Links

Gene Details

Gene
DPCD
Gene Name
deleted in primary ciliary dyskinesia homolog (mouse)
There are no transcripts associated with this gene.

Gene
MIR3158-1
Gene Name
microRNA 3158-1
There are no transcripts associated with this gene.

Gene
MIR3158-2
Gene Name
microRNA 3158-2
There are no transcripts associated with this gene.

Gene
POLL
Gene Name
polymerase (DNA) lambda
There are no transcripts associated with this gene.

View Full Product Details