Product Details
- SNP ID
-
rs7955866
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:4370383 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAAGGGGCGGCAGCCTTCCGGGCCC[A/G]TTCCCCCAGCGTGCGTGTTCACTCG
- Phenotype
-
MIM: 605380
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FGF23
PubMed Links
Gene Details
- Gene
- FGF23
- Gene Name
- fibroblast growth factor 23
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020638.2 |
862 |
Missense Mutation |
ACG,ATG |
T239M |
NP_065689.1 |
View Full Product Details