Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001345.4 | Intron | NP_001336.2 | |||
NM_201444.2 | Intron | NP_958852.1 | |||
NM_201445.1 | Intron | NP_958853.1 | |||
NM_201554.1 | Intron | NP_963848.1 | |||
XM_005268688.2 | Intron | XP_005268745.1 | |||
XM_005268689.2 | Intron | XP_005268746.1 | |||
XM_005268690.2 | Intron | XP_005268747.1 | |||
XM_011537991.2 | Intron | XP_011536293.1 | |||
XM_011537993.2 | Intron | XP_011536295.1 | |||
XM_011537995.2 | Intron | XP_011536297.1 | |||
XM_017018900.1 | Intron | XP_016874389.1 | |||
XM_017018901.1 | Intron | XP_016874390.1 | |||
XM_017018902.1 | Intron | XP_016874391.1 | |||
XM_017018903.1 | Intron | XP_016874392.1 | |||
XM_017018904.1 | Intron | XP_016874393.1 | |||
XM_017018905.1 | Intron | XP_016874394.1 | |||
XM_017018906.1 | Intron | XP_016874395.1 | |||
XM_017018907.1 | Intron | XP_016874396.1 | |||
XM_017018908.1 | Intron | XP_016874397.1 | |||
XM_017018909.1 | Intron | XP_016874398.1 |