Product Details

SNP ID
rs6070980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:59866447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAACTGTAGCATTCAGAATATA[G/T]GAATATGTAGCATTCAAAAGTTTTC
Phenotype
MIM: 604105
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SYCP2 PubMed Links

Gene Details

Gene
SYCP2
Gene Name
synaptonemal complex protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014258.3 Intron NP_055073.2
XM_011528487.2 Intron XP_011526789.1
XM_011528488.2 Intron XP_011526790.1
XM_011528489.2 Intron XP_011526791.1
XM_011528490.2 Intron XP_011526792.1
XM_011528493.2 Intron XP_011526795.1
XM_017027586.1 Intron XP_016883075.1
XM_017027587.1 Intron XP_016883076.1
XM_017027588.1 Intron XP_016883077.1
XM_017027589.1 Intron XP_016883078.1
XM_017027590.1 Intron XP_016883079.1
XM_017027591.1 Intron XP_016883080.1
XM_017027592.1 Intron XP_016883081.1

View Full Product Details