Product Details

SNP ID
rs139951223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151817281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGTTCATTCCATTTGAGTTTATT[A/T]AGGGTTAATGTCTACCTATAAGTCC
Phenotype
MIM: 600338
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AADAC PubMed Links

Gene Details

Gene
AADAC
Gene Name
arylacetamide deacetylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001086.2 Intron NP_001077.2
XM_005247104.4 Intron XP_005247161.1
Gene
AADACL2-AS1
Gene Name
AADACL2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details