Product Details

SNP ID
rs34895002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67675359 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTGCGGCGGTCGCCCTGCCCCTG[A/G]AAGGACTGGCGCAAGGCCTGCACCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf86 PubMed Links

Gene Details

Gene
C16orf86
Gene Name
chromosome 16 open reading frame 86
There are no transcripts associated with this gene.

Gene
ENKD1
Gene Name
enkurin domain containing 1
There are no transcripts associated with this gene.

Gene
GFOD2
Gene Name
glucose-fructose oxidoreductase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243650.1 992 Intron NP_001230579.1
NM_030819.3 992 Silent Mutation TTC,TTT F318F NP_110446.3
XM_006721288.3 992 Silent Mutation TTC,TTT F213F XP_006721351.1
XM_017023738.1 992 Silent Mutation TTC,TTT F318F XP_016879227.1

View Full Product Details