Product Details

SNP ID
rs3213190
Assay Type
Functionally tested
NCBI dbSNP Submissions
87
Location
Chr.1:235977874 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
AATAAGGCACTCTTGTCTTCATTTC[C/T]GTTCGATACAGTCAACTCCCAAGGT
Phenotype
MIM: 131390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NID1 PubMed Links

Gene Details

Gene
NID1
Gene Name
nidogen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002508.2 3839 Missense Mutation CAG,CGG Q1246R NP_002499.2
XM_011544195.2 3839 Missense Mutation CAG,CGG Q1204R XP_011542497.1

View Full Product Details