Product Details

SNP ID
rs61730329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29673166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGTTCACATCCAAAAGCCCCTG[G/T]ATGGACCTCTGGCTTCTGGCGATGG
Phenotype
MIM: 159465 MIM: 612192
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MOG PubMed Links

Gene Details

Gene
MOG
Gene Name
myelin oligodendrocyte glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008228.2 911 Intron NP_001008229.1
NM_001008229.2 911 Intron NP_001008230.1
NM_001170418.1 911 Intron NP_001163889.1
NM_002433.4 911 Intron NP_002424.3
NM_206809.3 911 Intron NP_996532.2
NM_206810.3 911 Intron NP_996533.2
NM_206811.3 911 Intron NP_996534.2
NM_206812.3 911 Intron NP_996535.2
NM_206814.5 911 Intron NP_996537.3
XM_005249131.3 911 Intron XP_005249188.1
Gene
ZFP57
Gene Name
ZFP57 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109809.2 911 Silent Mutation ATA,ATC I315I NP_001103279.2
XM_006715087.3 911 Silent Mutation ATA,ATC I243I XP_006715150.1
XM_011514570.2 911 Silent Mutation ATA,ATC I315I XP_011512872.1

View Full Product Details