Product Details

SNP ID
rs34383505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:128259461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCAAGTCTGTGAAGGGTTAATA[C/G]AGCTGAATCTGCAGCCTCATTCTGA
Phenotype
MIM: 612570
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBN2 PubMed Links

Gene Details

Gene
FBN2
Gene Name
fibrillin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001999.3 9335 Silent Mutation CTC,CTG L2911L NP_001990.2
XM_017009228.1 9335 Silent Mutation CTC,CTG L2860L XP_016864717.1

View Full Product Details