Product Details

SNP ID
rs115007276
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:32334866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAGAGAAAAATAAAGCCTGATT[A/G]GTACATATATTCCAGTTTTTCTTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf75 PubMed Links

Gene Details

Gene
C17orf75
Gene Name
chromosome 17 open reading frame 75
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022344.3 Intron NP_071739.2
XM_005258022.3 Intron XP_005258079.1
XM_017024940.1 Intron XP_016880429.1
XM_017024941.1 Intron XP_016880430.1

View Full Product Details