Product Details
- SNP ID
-
rs34027928
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:34320363 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGCTACACAAGAATCACCAACAT[A/C]CAATGTCCCAAGGAAGCTGTGATGT
- Phenotype
-
MIM: 602283
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CCL8
PubMed Links
Gene Details
- Gene
- CCL8
- Gene Name
- C-C motif chemokine ligand 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005623.2 |
626 |
Silent Mutation |
ATA,ATC |
I57I |
NP_005614.2 |
View Full Product Details