Product Details

SNP ID
rs45495899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52418122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATCACTAAAACAGAAGAGTATA[T/C]TTTAAAATTACCATTTATCAATAAT
Phenotype
MIM: 610903
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
VPS36 PubMed Links

Gene Details

Gene
VPS36
Gene Name
vacuolar protein sorting 36 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282168.1 Intron NP_001269097.1
NM_001282169.1 Intron NP_001269098.1
NM_016075.3 Intron NP_057159.2

View Full Product Details