Product Details

SNP ID
rs143475528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:95801728 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCGTCTCAGCAGGGGCTCCAGA[A/C]TTCCCCAGCAGGCGGCAGGTTTCCT
Phenotype
MIM: 601184 MIM: 605898
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC3 PubMed Links

Gene Details

Gene
DNAJC3
Gene Name
DnaJ heat shock protein family (Hsp40) member C3
There are no transcripts associated with this gene.

Gene
UGGT2
Gene Name
UDP-glucose glycoprotein glucosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020121.3 4819 UTR 3 NP_064506.3
XM_011521094.1 4819 Intron XP_011519396.1
XM_011521095.1 4819 Intron XP_011519397.1
XM_011521096.1 4819 Intron XP_011519398.1
XM_011521097.1 4819 UTR 3 XP_011519399.1
XM_011521098.1 4819 Intron XP_011519400.1
XM_011521099.2 4819 Intron XP_011519401.1
XM_011521100.2 4819 Intron XP_011519402.1
XM_011521102.2 4819 Intron XP_011519404.1
XM_011521103.2 4819 Intron XP_011519405.1
XM_017020662.1 4819 Intron XP_016876151.1
XM_017020663.1 4819 Intron XP_016876152.1

View Full Product Details