Product Details
- SNP ID
-
hCV25616446
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:49945330 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGCCCTTGTTTGGGACTATATTTT[C/G]CCTGTTGCAAGGTCCTCACTAACTT
- Phenotype
-
MIM: 612451
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RNF114
PubMed Links
Gene Details
- Gene
- RNF114
- Gene Name
- ring finger protein 114
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018683.3 |
|
Intron |
|
|
NP_061153.1 |
View Full Product Details