Product Details

SNP ID
rs28926211
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:95816718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAAATATTTCCCGTGATTCCAAA[C/G]TTTTCAAATCACTTTAAAACCTCCT
Phenotype
MIM: 600443
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GLRX PubMed Links

Gene Details

Gene
GLRX
Gene Name
glutaredoxin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001118890.1 Intron NP_001112362.1
NM_001243658.1 Intron NP_001230587.1
NM_001243659.1 Intron NP_001230588.1
NM_002064.2 Intron NP_002055.1

View Full Product Details