Product Details

SNP ID
rs10469471
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17251717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTCCAGTGTCCTGTTGCGAAGG[A/G]GAAGAGAGGGGGCTCACAGCCCCAG
Phenotype
MIM: 132880 MIM: 611810
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NR2F6 PubMed Links

Gene Details

Gene
NR2F6
Gene Name
nuclear receptor subfamily 2 group F member 6
There are no transcripts associated with this gene.

Gene
USHBP1
Gene Name
USH1 protein network component harmonin binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297703.1 Intron NP_001284632.1
NM_001321417.1 Intron NP_001308346.1
NM_031941.3 Intron NP_114147.2
XM_011528351.2 Intron XP_011526653.1

View Full Product Details