Product Details

SNP ID
rs977629997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:145937674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACGAGTTGAGTCTTATACCTGC[C/T]AGCATTGAATCAGAATGAGTCATTT
Phenotype
MIM: 615762 MIM: 613377
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRXCR2 PubMed Links

Gene Details

Gene
GRXCR2
Gene Name
glutaredoxin and cysteine rich domain containing 2
There are no transcripts associated with this gene.

Gene
SH3RF2
Gene Name
SH3 domain containing ring finger 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152550.3 Intron NP_689763.3

View Full Product Details