Product Details
- SNP ID
-
rs112359833
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:82276331 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTTTCATGTGAGGAAACTCCCTT[G/T]CATCAGATAAAAATGTGAGGGAGGC
- Phenotype
-
MIM: 603683
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP6AP1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111635232] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP6AP1L
- Gene Name
- ATPase H+ transporting accessory protein 1 like
There are no transcripts associated with this gene.
- Gene
- RPS23
- Gene Name
- ribosomal protein S23
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001025.4 |
|
Intron |
|
|
NP_001016.1 |
View Full Product Details