Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001253852.2 | 475 | Intron | NP_001240781.1 | ||
NM_001253853.2 | 475 | Intron | NP_001240782.1 | ||
NM_001308312.1 | 475 | Intron | NP_001295241.1 | ||
NM_006594.4 | 475 | Intron | NP_006585.2 | ||
XM_011540523.2 | 475 | Intron | XP_011538825.1 | ||
XM_011540524.2 | 475 | Intron | XP_011538826.1 | ||
XM_011540525.2 | 475 | Intron | XP_011538827.1 | ||
XM_011540528.2 | 475 | Intron | XP_011538830.1 | ||
XM_017000088.1 | 475 | Intron | XP_016855577.1 | ||
XM_017000089.1 | 475 | Intron | XP_016855578.1 | ||
XM_017000090.1 | 475 | Intron | XP_016855579.1 | ||
XM_017000091.1 | 475 | Intron | XP_016855580.1 | ||
XM_017000092.1 | 475 | Intron | XP_016855581.1 | ||
XM_017000093.1 | 475 | Intron | XP_016855582.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001319946.1 | 475 | UTR 5 | NP_001306875.1 | ||
NM_001319947.1 | 475 | UTR 5 | NP_001306876.1 | ||
NM_022836.3 | 475 | Missense Mutation | CAC,TAC | H61Y | NP_073747.1 |