Product Details

SNP ID
rs6032298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45553030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTTCCCAAAGATCCTCAAAGATG[A/G]GCCCAAGGAGCATCTGAGGTTCAAG
Phenotype
MIM: 609031
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPPIN PubMed Links

Gene Details

Gene
EPPIN
Gene Name
epididymal peptidase inhibitor
There are no transcripts associated with this gene.

Gene
EPPIN-WFDC6
Gene Name
EPPIN-WFDC6 readthrough
There are no transcripts associated with this gene.

Gene
WFDC8
Gene Name
WAP four-disulfide core domain 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130896.2 750 Intron NP_570966.2
NM_181510.2 750 Intron NP_852611.2
XM_017028119.1 750 Missense Mutation CCC,CTC P231L XP_016883608.1

View Full Product Details