Product Details
- SNP ID
-
rs3816544
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:51683561 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTATTAGTCAAGTCCAAAATTGAG[A/G]TAAAGCATATATTTACTGATAACAT
- Phenotype
-
MIM: 611796
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SCG3
PubMed Links
Gene Details
- Gene
- SCG3
- Gene Name
- secretogranin III
View Full Product Details