Product Details

SNP ID
rs143876260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:72816930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGGTTCTAACCCCAACAGCTGCC[C/T]GCCCAGGCGCCCTGCGGGACCCGGG
Phenotype
MIM: 602901
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNPO1 PubMed Links

Gene Details

Gene
TNPO1
Gene Name
transportin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002270.3 Intron NP_002261.3
NM_153188.2 Intron NP_694858.1
XM_005248500.2 Intron XP_005248557.1
XM_005248501.1 Intron XP_005248558.1
XM_017009458.1 Intron XP_016864947.1

View Full Product Details