Product Details

SNP ID
rs11542753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50959118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGCTAATCGTGGGCTGCGGTGG[A/G]CTCGGCTGTCCACTAGCGCAGTACT
Phenotype
MIM: 603503 MIM: 609277
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPM1 PubMed Links

Gene Details

Gene
DPM1
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317034.1 305 Intron NP_001303963.1
NM_001317035.1 305 Intron NP_001303964.1
NM_001317036.1 305 Intron NP_001303965.1
NM_003859.2 305 Intron NP_003850.1
XM_011529093.2 305 Intron XP_011527395.1
Gene
MOCS3
Gene Name
molybdenum cofactor synthesis 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014484.4 305 Silent Mutation GGA,GGG G92G NP_055299.1

View Full Product Details