Product Details

SNP ID
rs9783666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24095475 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCTTCTACCCCAGACAGACTCAA[G/T]CTCCCGTATCCATGCTCTGAGCTTT
Phenotype
MIM: 162080 MIM: 614095
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NRL PubMed Links

Gene Details

Gene
NRL
Gene Name
neural retina leucine zipper
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006177.3 776 Intron NP_006168.1
XM_005267708.4 776 Intron XP_005267765.1
XM_005267709.3 776 Intron XP_005267766.1
XM_005267710.3 776 Intron XP_005267767.1
XM_011536801.2 776 Intron XP_011535103.2
XM_011536802.1 776 UTR 5 XP_011535104.1
XM_011536804.2 776 Intron XP_011535106.1
XM_011536805.2 776 Intron XP_011535107.1
XM_011536806.2 776 Intron XP_011535108.2
XM_017021350.1 776 Intron XP_016876839.1
Gene
PCK2
Gene Name
phosphoenolpyruvate carboxykinase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018073.2 776 Intron NP_001018083.2
NM_001291556.1 776 Intron NP_001278485.1
NM_001308054.1 776 Intron NP_001294983.1
NM_004563.3 776 Intron NP_004554.3
XM_006720158.2 776 Intron XP_006720221.1

View Full Product Details