Product Details

SNP ID
rs16957485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43197503 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGGCAGGTGCTAGTTCTGTCCCA[C/G]GTTCATTGCTCTGCTGGTCCCAGTG
Phenotype
MIM: 607089 MIM: 177070
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCNDBP1 PubMed Links

Gene Details

Gene
CCNDBP1
Gene Name
cyclin D1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012142.4 Intron NP_036274.3
XM_006720448.1 Intron XP_006720511.1
Gene
EPB42
Gene Name
erythrocyte membrane protein band 4.2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000119.2 Intron NP_000110.2
NM_001114134.1 Intron NP_001107606.1
XM_005254225.1 Intron XP_005254282.1
XM_011521349.2 Intron XP_011519651.1
XM_011521350.2 Intron XP_011519652.1
XM_011521351.2 Intron XP_011519653.1
XM_011521352.2 Intron XP_011519654.1
XM_011521353.2 Intron XP_011519655.1
XM_011521354.2 Intron XP_011519656.1

View Full Product Details