Product Details
- SNP ID
-
rs112450811
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:24631435 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCAGCCTCCGTCATGTCCCCCAG[A/G]TCCTGAGATCGGAGGTATGTGGGAC
- Phenotype
-
MIM: 123910
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GZMB
PubMed Links
Gene Details
- Gene
- GZMB
- Gene Name
- granzyme B
View Full Product Details