Product Details

SNP ID
hCV25624228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:160190731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACCTCTTCTGGACCAGGAGAGCC[A/C]ACCCAGATCCCACTACCTCCATGAG
Phenotype
MIM: 607321 MIM: 114250
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATP1A4 PubMed Links

Gene Details

Gene
ATP1A4
Gene Name
ATPase Na+/K+ transporting subunit alpha 4
There are no transcripts associated with this gene.

Gene
CASQ1
Gene Name
calsequestrin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001231.4 237 UTR 5 NP_001222.3

View Full Product Details