Product Details
- SNP ID
-
rs752118047
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:68932532 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAATGTTTTGCAAATTTTGGTACCT[C/G]TTATGCTAAGTGGAAAATGAGATTC
- Phenotype
-
MIM: 616382
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
UGT2A3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115826767] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UGT2A3
- Gene Name
- UDP glucuronosyltransferase family 2 member A3
View Full Product Details