Product Details

SNP ID
hCV25625156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:114942914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTAAAGAAGCCCTTAAATGGTA[C/G]AAGGCCTACAAGGCTGTTAGAAATA
Phenotype
MIM: 610726
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRUB1 PubMed Links

Gene Details

Gene
TRUB1
Gene Name
TruB pseudouridine synthase family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139169.4 Intron NP_631908.1

View Full Product Details