Product Details

SNP ID
rs34393297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:114942666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGAAGCTGGAATGCCTTCTCCAG[A/C]ATGGACCAAGAGGAAAAAGCAGACT
Phenotype
MIM: 610726
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TRUB1 PubMed Links

Gene Details

Gene
TRUB1
Gene Name
TruB pseudouridine synthase family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139169.4 369 Missense Mutation GAA,GCA E103A NP_631908.1

View Full Product Details