Product Details

SNP ID
rs115865042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:26599444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGAAAAATGGGGGAAATAATT[A/G]AACATGAAGAAGGGTTGGGGCAAGG
Phenotype
MIM: 123631 MIM: 600929 MIM: 603126
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRYBA4 PubMed Links

Gene Details

Gene
CRYBA4
Gene Name
crystallin beta A4
There are no transcripts associated with this gene.

Gene
CRYBB1
Gene Name
crystallin beta B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001887.3 875 UTR 3 NP_001878.1
XM_011529899.2 875 UTR 3 XP_011528201.1
Gene
TPST2
Gene Name
tyrosylprotein sulfotransferase 2
There are no transcripts associated with this gene.

View Full Product Details