Product Details
- SNP ID
-
rs61730833
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:55105493 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAACAGATGTCTGTGAAAAAATTAT[C/T]GGAGGAAATGAAGTAACTCCTCATT
- Phenotype
-
MIM: 140050
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDC20B
PubMed Links
Gene Details
- Gene
- CDC20B
- Gene Name
- cell division cycle 20B
There are no transcripts associated with this gene.
- Gene
- GZMA
- Gene Name
- granzyme A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006144.3 |
127 |
Silent Mutation |
ATC,ATT |
I30I |
NP_006135.1 |
View Full Product Details