Product Details

SNP ID
rs61730859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:49185844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTACAGCTGTGGAAACCTGGGG[C/T]GCCGGGTAAATAGAGAACTCCAGCT
Phenotype
MIM: 602529
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TUBA1A PubMed Links

Gene Details

Gene
TUBA1A
Gene Name
tubulin alpha 1a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270399.1 955 Silent Mutation GCA,GCG A174A NP_001257328.1
NM_001270400.1 955 Silent Mutation GCA,GCG A139A NP_001257329.1
NM_006009.3 955 Silent Mutation GCA,GCG A174A NP_006000.2

View Full Product Details