Product Details

SNP ID
rs111502709
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141625959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCCTTCAAACCAGGTCATTCTG[C/G]AATCTGGTGAGAATGGCCTTCCTGT
Phenotype
MIM: 602121 MIM: 605166
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DIAPH1 PubMed Links

Gene Details

Gene
DIAPH1
Gene Name
diaphanous related formin 1
There are no transcripts associated with this gene.

Gene
HDAC3
Gene Name
histone deacetylase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003883.3 Intron NP_003874.2
XM_011537697.2 Intron XP_011535999.1

View Full Product Details