Product Details

SNP ID
rs72624917
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49024852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGGTCACAGCAGAGATGAGACT[A/G]TCAGTGCAGGATTAGGGTGGGGTAA
Phenotype
MIM: 146691 MIM: 615150 MIM: 612911 MIM: 606031
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DALRD3 PubMed Links

Gene Details

Gene
DALRD3
Gene Name
DALR anticodon binding domain containing 3
There are no transcripts associated with this gene.

Gene
IMPDH2
Gene Name
inosine monophosphate dehydrogenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000884.2 Intron NP_000875.2
XM_006713128.3 Intron XP_006713191.1
XM_017006349.1 Intron XP_016861838.1
XM_017006350.1 Intron XP_016861839.1
Gene
MIR191
Gene Name
microRNA 191
There are no transcripts associated with this gene.

Gene
MIR425
Gene Name
microRNA 425
There are no transcripts associated with this gene.

Gene
NDUFAF3
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 3
There are no transcripts associated with this gene.

Gene
QRICH1
Gene Name
glutamine rich 1
There are no transcripts associated with this gene.

Gene
WDR6
Gene Name
WD repeat domain 6
There are no transcripts associated with this gene.

View Full Product Details