Product Details
- SNP ID
-
rs1805088
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:46008985 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGGTGCTGGGCTGCTGCTTTGCTG[C/T]CCCCAGACAGCGCCAGTCCACCCTT
- Phenotype
-
MIM: 120361
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MMP9
PubMed Links
Gene Details
- Gene
- MMP9
- Gene Name
- matrix metallopeptidase 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004994.2 |
78 |
Missense Mutation |
GCC,GTC |
A20V |
NP_004985.2 |
View Full Product Details