Product Details

SNP ID
rs78711713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42797584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGAAATACACATCATGACTTC[A/G]GTATTAAGAACCGTAGCGATCCTGA
Phenotype
MIM: 612439
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARFGAP3 PubMed Links

Gene Details

Gene
ARFGAP3
Gene Name
ADP ribosylation factor GTPase activating protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142293.1 1775 UTR 3 NP_001135765.1
NM_014570.4 1775 UTR 3 NP_055385.3
XM_005261525.3 1775 UTR 3 XP_005261582.1

View Full Product Details