Product Details

SNP ID
hCV25630904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32179290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGAGAGGGCACGAAAATTTGTA[C/T]GTGTTTGTTGGGGAGAGGAGAAAGG
Phenotype
MIM: 600214 MIM: 603099 MIM: 176311 MIM: 602677
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGER PubMed Links

Gene Details

Gene
AGER
Gene Name
advanced glycosylation end product-specific receptor
There are no transcripts associated with this gene.

Gene
AGPAT1
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 1
There are no transcripts associated with this gene.

Gene
MIR6721
Gene Name
microRNA 6721
There are no transcripts associated with this gene.

Gene
MIR6833
Gene Name
microRNA 6833
There are no transcripts associated with this gene.

Gene
PBX2
Gene Name
PBX homeobox 2
There are no transcripts associated with this gene.

Gene
RNF5
Gene Name
ring finger protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006913.3 Intron NP_008844.1

View Full Product Details