Product Details

SNP ID
rs12193672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35737365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGGCCCAAAGGTGATGCCTCCA[A/G]GTTCCTCTTTCCTTTGTCTTTCTCT
Phenotype
MIM: 602623
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARMC12 PubMed Links

Gene Details

Gene
ARMC12
Gene Name
armadillo repeat containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286574.1 Intron NP_001273503.1
NM_001286576.1 Intron NP_001273505.1
NM_145028.4 Intron NP_659465.2
XM_011514381.2 Intron XP_011512683.1
XM_011514382.2 Intron XP_011512684.1
XM_017010435.1 Intron XP_016865924.1
Gene
FKBP5
Gene Name
FK506 binding protein 5
There are no transcripts associated with this gene.

Gene
LOC285847
Gene Name
uncharacterized LOC285847
There are no transcripts associated with this gene.

View Full Product Details