Product Details

SNP ID
rs779332895
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:132682132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAACTCTTAAAATGAGAATATAA[A/C]CTCTTACCAATTCAAAATGACCTTA
Phenotype
MIM: 608002 MIM: 610552
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NPHP3 PubMed Links

Gene Details

Gene
NPHP3
Gene Name
nephronophthisis 3 (adolescent)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153240.4 Intron NP_694972.3
Gene
NPHP3-ACAD11
Gene Name
NPHP3-ACAD11 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
UBA5
Gene Name
ubiquitin like modifier activating enzyme 5
There are no transcripts associated with this gene.

View Full Product Details