Product Details

SNP ID
rs561600265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19180452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCCCAGCACACACCAGCAGCAG[C/T]CTTGCTGTCCAGGGGCCAGGCCCAG
Phenotype
MIM: 601273 MIM: 190315
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLTCL1 PubMed Links

Gene Details

Gene
CLTCL1
Gene Name
clathrin heavy chain like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001835.3 Intron NP_001826.3
NM_007098.3 Intron NP_009029.3
XM_011530401.2 Intron XP_011528703.1
XM_017028953.1 Intron XP_016884442.1
XM_017028954.1 Intron XP_016884443.1
XM_017028955.1 Intron XP_016884444.1
XM_017028956.1 Intron XP_016884445.1
XM_017028957.1 Intron XP_016884446.1
Gene
LINC01311
Gene Name
long intergenic non-protein coding RNA 1311
There are no transcripts associated with this gene.

Gene
SLC25A1
Gene Name
solute carrier family 25 member 1
There are no transcripts associated with this gene.

View Full Product Details